5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole ≥96.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
5-Oxindoleboronic acid Pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-2-one, 2-(Oxindole-5-yl)-4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
T3512-1G
T3512-5G
TCIAT3512-1GEA
119
EUR
TCIAT3512-1G
TCIAT3512-5G
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole ≥96.0% (by GC, titration analysis)
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole
Formula:
C₁₄H₁₈BNO₃ MW: 259,11 g/mol |
Número MDL:
MFCD08669935 Núm. CAS: 837392-64-0 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 232.0 to 236.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...