Tris(1,10-phenanthroline)cobalt(III) tris(hexafluorophosphate) ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Atención

T3554-1G T3554-5G
TCIAT3554-1GEA 111 EUR
TCIAT3554-1G TCIAT3554-5G
Tris(1,10-phenanthroline)cobalt(III) tris(hexafluorophosphate) ≥97.0% (by titrimetric analysis)
Tris(1,10-phenanthroline)cobalt(III) tris(hexafluorophosphate)
Formula: C₃₆H₂₄CoF₁₈N₆P₃
MW: 1034,46 g/mol
Número MDL: MFCD30749311
Núm. CAS: 28277-59-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fae11319c6026c7664bf2823613ef182347cacd&vid=2fae11319c6026c7664bf2823613ef182347cacd&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Chelometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR