Sinónimos:
2-[4-(Methanesulphonylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-(Methylsulphonyl)-4-(4,4,5,5-tetraMethyl-1,3,2-dioxaborolan-2-yl)benzenamine, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanesulphonamide, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-phenyl)methanesulphonamide, 4-(Methanesulphonylamino)phenylboronic acid Pinacol ester, 4-Methanesulphonylaminophenylboronic acid, pinacol ester, N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulphonamide, 4-(Methylsulphonamido)phenylboronic acid pinacol ester
T3557-1G
T3557-5G
TCIAT3557-1GEA
39
EUR
TCIAT3557-1G
TCIAT3557-5G
4-(Methanesulphonamido)phenylboronic acid pinacol ester ≥98.0% (by titrimetric analysis)
4-(Methanesulphonamido)phenylboronic acid pinacol ester
Formula:
C₁₃H₂₀BNO₄S
|
Número MDL:
MFCD05663876
Núm. CAS:
616880-14-9
|
New Product
Limited Time Sale
Select items below are currently on sale
Order Now