Tigecycline ≥98.0% (by HPLC)
Proveedor: TCI
|
Peligro
|
Sinónimos:
9-tert-Butylglycylamidominocycline, (4S,4aS,5aR,12aS)-9-(2-(tert-Butylamino)acetamido)-4,7-bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide
T3589-100MG
T3589-500MG
TCIAT3589-100MGEA
62
EUR
TCIAT3589-100MG
TCIAT3589-500MG
Tigecycline ≥98.0% (by HPLC)
Tigecycline
Formula:
C₂₉H₃₉N₅O₈ MW: 585,658 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00935753 Núm. CAS: 220620-09-7 UN: 3077 ADR: 9,III Índice Merck: 14,09432 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Specific rotation [a]20/D | -189 to -208 deg (C=0.15, methanol) |
Specific rotation (value) | -208 to -189 deg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...