Trimethyl(nonafluorobutyl)silane ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
T3594-1G
T3594-5G
TCIAT3594-1GEA
92
EUR
TCIAT3594-1G
TCIAT3594-5G
Trimethyl(nonafluorobutyl)silane ≥98.0% (por GC)
Trimethyl(nonafluorobutyl)silane
Formula:
C₇H₉F₉Si Pto. Ebullición: 100°C |
Número MDL:
MFCD20489405 Núm. CAS: 204316-01-8 UN: 1993 ADR: 3,II |
|
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...