Trimethyl(tridecafluorohexyl)silane ≥97.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
Trimethyl(perfluorohexyl)silane
T3595-1G
T3595-5G
TCIAT3595-1GEA
84
EUR
TCIAT3595-1G
TCIAT3595-5G
Trimethyl(tridecafluorohexyl)silane ≥97.0% (por GC)
Trimethyl(tridecafluorohexyl)silane
Formula:
C₉H₉F₁₃Si MW: 392,24 g/mol |
Número MDL:
MFCD20489406 Núm. CAS: 135841-49-5 UN: 1993 ADR: 3,III |
Resultados test especificaciones
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...