TWS119 ≥95.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-[[6-(3-Aminophenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]oxy]phenol, 3-((6-(3-Aminophenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl)oxy)phenol
T3605-5MG
T3605-25MG
TCIAT3605-5MGEA
54
EUR
TCIAT3605-5MG
TCIAT3605-25MG
TWS119 ≥95.0% (by HPLC)
TWS119
This product is unavailable in the U.S.
Formula:
C₁₈H₁₄N₄O₂ MW: 318,34 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD09037535 Núm. CAS: 601514-19-6 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Melting point | 247.0 to 251.0 deg-C |
Water (Vaporization methoed) | max. 5.0 % |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...