Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
N-[6-(Dimethylamino)-9-[2-(methoxycarbonyl)phenyl]-3H-xanthen-3-ylidene]-N-methylmethanaminium Perchlorate, Tetramethylrhodamine methyll ester Perchlorate, 3,6-Bis(dimethylamino)-9-(2-(methoxycarbonyl)phenyl)xanthylium perchlorate, Tetramethylrhodamine Methyl Ester Perchlorate
T3608-10MG
T3608-50MG
TCIAT3608-10MGEA
76
EUR
TCIAT3608-10MG
TCIAT3608-50MG
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥98.0% (by HPLC)
Tetramethylrhodamine methyl ester perchlorate (TMRM)
Formula:
C₂₅H₂₅N₂O₃·ClO₄ MW: 500,93 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00274460 Núm. CAS: 115532-50-8 UN: 1479 ADR: 5.1,III |
Resultados test especificaciones
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 5.20 to 6.00 % |
Molar extinction coefficient | min. 100000 (MeOH, 497.0 to 553.0nm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...