3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9-(p-tolyl)-9H-carbazole ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
9-(p-Tolyl)-9H-carbazole-3-boronic acid Pinacol ester, 2-[9-(p-Tolyl)-9H-carbazol-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
T3612-200MG
T3612-1G
TCIAT3612-200MGEA
58
EUR
TCIAT3612-200MG
TCIAT3612-1G
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9-(p-tolyl)-9H-carbazole ≥98.0% (by HPLC)
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9-(p-tolyl)-9H-carbazole
Formula:
C₂₅H₂₆BNO₂ MW: 383,30 g/mol |
Núm. CAS:
1345614-94-9 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 164.0 to 168.0 deg-C |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...