3,4,5-Tris(dodecyloxy)benzaldehyde ≥98.0% (por GC)
Proveedor: TCI
T3624-200MG
T3624-1G
TCIAT3624-200MGEA
52
EUR
TCIAT3624-200MG
TCIAT3624-1G
3,4,5-Tris(dodecyloxy)benzaldehyde ≥98.0% (por GC)
3,4,5-Tris(dodecyloxy)benzaldehyde
Formula:
C₄₃H₇₈O₄ MW: 659,09 g/mol |
Número MDL:
MFCD29917550 Núm. CAS: 117241-32-4 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Acid Value | max. 2.0 |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...