1,1'-Thiocarbonyldi-2(1H)-pyridone ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
1,1'-Thiocarbonylbis(pyridin-2(1H)-one)
T3644-1G
T3644-5G
TCIAT3644-1GEA
29
EUR
TCIAT3644-1G
TCIAT3644-5G
1,1'-Thiocarbonyldi-2(1H)-pyridone ≥98.0% (by HPLC, total nitrogen)
1,1'-Thiocarbonyldi-2(1H)-pyridone
Formula:
C₁₁H₈N₂O₂S MW: 232,26 g/mol |
Número MDL:
MFCD00075238 Núm. CAS: 102368-13-8 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 156.0 to 160.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...