4-(Trimethylsilylethynyl)aniline ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-((Trimethylsilyl)ethynyl)aniline, 4-[(Trimethylsilyl)ethynyl]aniline
T3702-1G
T3702-250MG
TCIAT3702-1GEA
150
EUR
TCIAT3702-1G
TCIAT3702-250MG
4-(Trimethylsilylethynyl)aniline ≥98.0% (by GC, titration analysis)
4-(Trimethylsilylethynyl)aniline
Formula:
C₁₁H₁₅NSi MW: 189,33 g/mol |
Número MDL:
MFCD03695479 Núm. CAS: 75867-39-9 |
Resultados test especificaciones
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...