Tris(2-phenylpyridinato)iridium(III) ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
Tris(2-phenylpyridine)iridium, Ir(ppy)3
T3716-200MG
T3716-1G
TCIAT3716-200MGEA
433
EUR
TCIAT3716-200MG
TCIAT3716-1G
Tris(2-phenylpyridinato)iridium(III) ≥98.0% (by HPLC)
Tris(2-phenylpyridinato)iridium(III)
Formula:
C₃₃H₂₄IrN₃ MW: 654,79 g/mol |
Número MDL:
MFCD04039766 Núm. CAS: 94928-86-6 |
|
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystaline |
Purity (HPLC) | min. 98.0 % (sum of fac and mer isomers) |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...