1-(Trifluoromethyl)cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

T3765-200MG T3765-1G
TCIAT3765-200MGEA 22 EUR
TCIAT3765-200MG TCIAT3765-1G
1-(Trifluoromethyl)cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(Trifluoromethyl)cyclopropanecarboxylic acid
Formula: C₅H₅F₃O₂
Pto. Ebullición: 184°C
Número MDL: MFCD03093070
Núm. CAS: 277756-46-4
UN: 3261
ADR: 8,II

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR