1,3,5-Triazinane-2,4-dione ≥97.0% (by total nitrogen basis)
Proveedor: TCI
T3800-1G
T3800-5G
TCIAT3800-1GEA
76
EUR
TCIAT3800-1G
TCIAT3800-5G
1,3,5-Triazinane-2,4-dione ≥97.0% (by total nitrogen basis)
1,3,5-Triazinane-2,4-dione
Formula:
C₃H₅N₃O₂ |
Número MDL:
MFCD00085440 Núm. CAS: 27032-78-6 EINECS: 248-178-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...