3-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
2-[3-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hydroxymethylphenylboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl alcohol, (3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol, 3-(Hydroxymethyl)phenylboronic acid Pinacol ester, 3-(Hydroxymethyl)benzeneboronic acid pinacol ester
T3830-1G
T3830-5G
TCIAT3830-1GEA
18
EUR
TCIAT3830-1G
TCIAT3830-5G
3-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-(Hydroxymethyl)phenylboronic acid pinacol ester
Formula:
C₁₃H₁₉BO₃ MW: 234.10 g/mol |
Número MDL:
MFCD09266196 Núm. CAS: 443776-76-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...