4-(Trifluoromethyl)aniline hydrobromide ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Aminobenzotrifluoride hydrobromide, 4-(Trifluoromethyl)benzenamine hydrobromide, 4-(Trifluoromethyl)anilinium bromide
T3834-1G
T3834-5G
TCIAT3834-1GEA
66
EUR
TCIAT3834-1G
TCIAT3834-5G
4-(Trifluoromethyl)aniline hydrobromide ≥98.0% (by HPLC)
4-(Trifluoromethyl)aniline hydrobromide
Formula:
C₇H₆F₃N·HBr |
Núm. CAS:
148819-81-2 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...