4-(Trifluoromethyl)benzylamine hydrobromide ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
T3837-1G
T3837-5G
TCIAT3837-1GEA
26
EUR
TCIAT3837-1G
TCIAT3837-5G
4-(Trifluoromethyl)benzylamine hydrobromide ≥98.0% (by HPLC)
4-(Trifluoromethyl)benzylamine hydrobromide
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...