4-(Trifluoromethyl)benzylamine Hydroiodide ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-(Trifluoromethyl)benzylammonium iodide, 4-Aminomethylbenzotrifluoride Hydroiodide, [4-(Trifluoromethyl)phenyl]methanamine Hydroiodide
T3838-1G
T3838-5G
TCIAT3838-1GEA
28
EUR
TCIAT3838-1G
TCIAT3838-5G
4-(Trifluoromethyl)benzylamine Hydroiodide ≥98.0% (by HPLC)
4-(Trifluoromethyl)benzylamine Hydroiodide
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...