Tetrahydro-2H-pyran-4-carbonyl chloride ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

T3890-1G T3890-5G
TCIAT3890-1GEA 41 EUR
TCIAT3890-1G TCIAT3890-5G
Tetrahydro-2H-pyran-4-carbonyl chloride ≥97.0% (by GC, titration analysis)
Tetrahydro-2H-pyran-4-carbonyl chloride
Formula: C₆H₉ClO₂
Pto. Ebullición: 85°C
Número MDL: MFCD06200863
Núm. CAS: 40191-32-0
UN: 3265
ADR: 8,II

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Specific gravity (20/20) 1.2010 to 1.2050


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR