Undecylphosphonic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

U0088-1G U0088-5G
TCIAU0088-1GEA 165 EUR
TCIAU0088-1G TCIAU0088-5G
Undecylphosphonic acid ≥98.0% (by titrimetric analysis)
Undecylphosphonic acid
Formula: C₁₁H₂₅O₃P
Número MDL: MFCD00015831
Núm. CAS: 5137-69-9
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15a1d34d4280df72c405f7c2a17c8050b41fb643&vid=15a1d34d4280df72c405f7c2a17c8050b41fb643&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR