Sultamicillin Tosylate dihydrate ≥98.0% (by GC, titration analysis)

Proveedor: TCI

Sinónimos: Sultamicillin Tosilate

U0097-1G U0097-250MG
TCIAU0097-1GEA 424 EUR
TCIAU0097-1G TCIAU0097-250MG
Sultamicillin Tosylate dihydrate ≥98.0% (by GC, titration analysis)
Sultamicillin Tosylate dihydrate
Formula: C₂₅H₃₀N₄O₉S₂·C₇H₈O₃S·2H₂O
MW: 766,85 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD01682151
Núm. CAS: 83105-70-8
Índice Merck: 14,08992

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +168 to +186 deg (C=0.5, water/CH3CN 3:2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR