Vincamine ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Vincamine base, Methyl (41S,12S,13aS)-13a-ethyl-12-hydroxy-2,3,41,5,6,12,13,13a-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate
V0061-1G
V0061-5G
TCIAV0061-1GEA
76
EUR
TCIAV0061-1G
TCIAV0061-5G
Vincamine ≥98.0% (by titrimetric analysis)
Vincamine
Formula:
C₂₁H₂₆N₂O₃ MW: 354,45 g/mol |
Número MDL:
MFCD00078054 Núm. CAS: 1617-90-9 EINECS: 216-576-3 Índice Merck: 14,09983 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +42.0 to +44.0 deg (C=1, Pyridine) |
Solubility in hot DMF | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...