D-Valine methyl ester hydrochloride ≥98.0% (by total nitrogen basis)

Proveedor: TCI

Sinónimos: H-D-Val-OMe.HCl, H-D-Val-OMe·HCl

V0094-1G V0094-5G
TCIAV0094-1GEA 17 EUR
TCIAV0094-1G TCIAV0094-5G
D-Valine methyl ester hydrochloride ≥98.0% (by total nitrogen basis)
D-Valine methyl ester hydrochloride
Formula: C₆H₁₃NO₂
MW: 167.633 g/mol
Número MDL: MFCD00237309
Núm. CAS: 7146-15-8
EINECS: 230-454-7

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -13.0 to -17.0 deg (C=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR