Valnemulin hydrochloride ≥98.0% (by HPLC)
Proveedor: TCI
V0144-50MG
V0144-250MG
TCIAV0144-50MGEA
120
EUR
TCIAV0144-50MG
TCIAV0144-250MG
Valnemulin hydrochloride ≥98.0% (by HPLC)
Valnemulin hydrochloride
Formula:
C₃₁H₅₂N₂O₅S·HCl Temperatura de almacenaje: Congelador |
Número MDL:
MFCD08458958 Núm. CAS: 133868-46-9 Índice Merck: 14,09911 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 98.0 area% |
Elemental analysis (Carbon) | 59.00 to 64.00 % |
Elemental analysis (Nitrogen) | 4.00 to 5.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...