Valganciclovir hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: 2-[(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)methoxy]-3-hydroxypropyl (2S)-2-amino-3-methylbutanoate hydrochloride hydrate, Valcyte, Valganciclovir HCl, (2S)-2-((2-Amino-6-oxo-1H-purin-9(6H)-yl)methoxy)-3-hydroxypropyl 2-amino-3-methylbutanoate hydrochloride

V0158-1G V0158-250MG
TCIAV0158-1GEA 106 EUR
TCIAV0158-1G TCIAV0158-250MG
Valganciclovir hydrochloride ≥98.0% (by titrimetric analysis)
Valganciclovir hydrochloride
Formula: C₁₄H₂₂N₆O₅·HCl
MW: 390,83 g/mol
Número MDL: MFCD08460118
Núm. CAS: 175865-59-5
Índice Merck: 14,09907

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 % (calcd.on anh.substance)
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR