[Arg 8]-Vasopressin Acetate ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
Argipressin Acetate, AVP, beta-Hypophamine
V0159-25MG
V0159-5MG
TCIAV0159-25MGEA
639
EUR
TCIAV0159-25MG
TCIAV0159-5MG
[Arg 8]-Vasopressin Acetate ≥98.0% (by HPLC)
[Arg 8]-Vasopressin Acetate
Formula:
C₄₆H₆₅N₁₅O₁₂S₂·xC₂H₄O₂ Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00133939 Núm. CAS: 129979-57-3 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...