WHI-P154 ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
2-Bromo-4-[(6,7-dimethoxy-4-quinazolinyl)amino]phenol, 2-Bromo-4-((6,7-dimethoxyquinazolin-4-yl)amino)phenol
W0013-10MG
W0013-50MG
TCIAW0013-10MGEA
63
EUR
TCIAW0013-10MG
TCIAW0013-50MG
WHI-P154 ≥98.0% (by HPLC)
WHI-P154
Formula:
C₁₆H₁₄BrN₃O₃ MW: 376,2 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD02179366 Núm. CAS: 211555-04-3 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 228.0 to 232.0 deg-C |
Elemental analysis (Nitrogen) | 10.50 to 11.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...