Yttrium(III) isopropoxide ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Y0009-200MG Y0009-1G
TCIAY0009-200MGEA 72 EUR
TCIAY0009-200MG TCIAY0009-1G
Yttrium(III) isopropoxide ≥98.0% (by titrimetric analysis)
Yttrium(III) isopropoxide
Formula: C₉H₂₁O₃Y
Número MDL: MFCD00015641
Núm. CAS: 2172-12-5
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2210a721c8f77acd0fc22262e88296f59f3d6a12&vid=2210a721c8f77acd0fc22262e88296f59f3d6a12&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR