Zolmitriptan ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Z0024-200MG Z0024-1G
TCIAZ0024-200MGEA 29 EUR
TCIAZ0024-200MG TCIAZ0024-1G
Zolmitriptan ≥98.0% (by HPLC, titration analysis)
Zolmitriptan
Formula: C₁₆H₂₁N₃O₂
Temperatura de almacenaje: Nevera
Número MDL: MFCD00871503
Núm. CAS: 139264-17-8
Índice Merck: 14,10189
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bc4d4b78acd1cf933cd42cdf7ae201118c3618b&vid=9bc4d4b78acd1cf933cd42cdf7ae201118c3618b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C
Specific rotation [a]20/D -3.0 to -5.0 deg (C=1、MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR