Zinc(II) Bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: Zinc bis(trifluoromethylsulphonyl)imide, Bis(trifluoromethanesulphonyl)imide Zinc(II) Salt, Zinc(II) Triflimide

Z0027-1G Z0027-5G
TCIAZ0027-1GEA 62 EUR
TCIAZ0027-1G TCIAZ0027-5G
Zinc(II) Bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Zinc(II) Bis(trifluoromethanesulphonyl)imide
Formula: C₄F₁₂N₂O₈S₄Zn
MW: 625.65 g/mol
Número MDL: MFCD16621474
Núm. CAS: 168106-25-0
UN: 1759
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecd3ea022945e4f8d4aa085f92cf8b6405de8372&vid=ecd3ea022945e4f8d4aa085f92cf8b6405de8372&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR