Ziprasidone hydrochloride monohydrate ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 5-[2-[4-(1,2-Benzisothiazol-3-yl)-1-piperazinyl]ethyl]-6-chloro-1,3-dihydro-2H-indol-2-one Hydrochloride

Z0032-50MG Z0032-250MG
TCIAZ0032-50MGEA 27 EUR
TCIAZ0032-50MG TCIAZ0032-250MG
Ziprasidone hydrochloride monohydrate ≥98.0% (by HPLC)
Ziprasidone hydrochloride monohydrate
Formula: C₂₁H₂₁ClN₄OS·HCl·H₂O
Número MDL: MFCD00921885
Núm. CAS: 138982-67-9
Índice Merck: 14,10171
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8445912cfaddbaef804c3b96b7175a8e95c4dee&vid=e8445912cfaddbaef804c3b96b7175a8e95c4dee&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 % (calcd.on anh.substance)
Water 3.7 to 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR