PIPES free acid, white powder

Proveedor: MP Biomedicals

Sinónimos: Ácido 2,2'-(piperazina-1,4-diil)bis(etanosulfonico)

0219025710 0219025791 0219025780 0219025790
ICNA0219025710EA 33.9 EUR
ICNA0219025710 ICNA0219025791 ICNA0219025780 ICNA0219025790
PIPES free acid, white powder
PIPES

PIPES is a zwitterionic N-substituted aminosulfonic acid "Good" buffer. Buffers can be made by adding a solution of base to PIPES free acid, titrating to the appropriate pH, or by mixing equimolar solutions of the sesquisodium salt and the disodium salt or dipotassium salt, titrating to the appropriate pH.


Zwitterionic Buffer, Effective buffering range: 6,1 - 7,5 (Merck Index reports 6,0 to 8,5 ), ΔpK/ΔT: 0,0085, pKa: 6,8 at 25 °C ; 6,66 at 37 °C


PIPES is used to buffer glutaraldehyde fixative solutions when fixing plant and/or animal tissue tissue samples. This will help prevent the loss of lipids. It is used to buffer glutaraldehyde and formaldehyde when fixing fungal zoospores for fluorescence microscopy and electron microscopy.


PIPES free acid is not very water soluble (approximately 1 g/l at 100 °C) but will solubilise in 0,5 M NaOH; PIPES salts are water soluble (usually resulting in clear, colourless solutions).

Formula: C₈H₁₈N₂O₆S₂
Pto. Ebullición: 499 °C (1013 hPa)
Pto. de fusión: 300 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00006159
Núm. CAS: 5625-37-6
EINECS: 227-057-6
Índice Merck: 12,07633
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5192468b680c7bea85a31a131811618356a531b9&vid=ef0e3d488c68ea7f6f583fede6ebf7d2196392b7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Identity Test Passes
FTIR Conforms to Standard
UV/Vis: OD260 (0,1 M in 0,2 M NaOH) ≤0,05
UV/Vis: OD280 (0,1 M in 0,2 M NaOH) ≤0,05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR