Chemical resistant lab coat, AlphaTec® 1500 PLUS, Model 208
Proveedor: Ansell
WH15B-00208-06 WH15B-00208-05 WH15B-00208-03 WH15B-00208-04
ANSEWH15B-00208-06EA 323 EUR
ANSEWH15B-00208-06 ANSEWH15B-00208-05 ANSEWH15B-00208-03 ANSEWH15B-00208-04
Chemical resistant lab coat, AlphaTec® 1500 PLUS, Model 208
Prendas desechables
Breathable, anti-static SMS material.
  • Air and water vapour permeable (breathable)
  • Silicone-free
  • Optimised body fit improves wearer comfort and safety
Short length lab coats feature a collar, elasticated cuffs, studded front fastening, left breast and right thigh pockets. Garments are manufactured from SMS fabric with anti-static treatment. Bound seams.
Recommended for unpacking of bulk goods, loading and unloading of proces equipment, production line support and maintenance, and component assembly and inspection.
Certificaciones: Type PB [6]);More Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Género
Aplicaciones de vestimenta Chemical protection