Povidona iodada 10%
Proveedor: Alcoholes Montplet
Sinónimos:
Polyvinylpyrrolidone-iodine complex
PIMPBT066
PIMPBT068
ALCHPIMPBT066EA
4.1
EUR
ALCHPIMPBT066
ALCHPIMPBT068
Povidona iodada 10%
Povidona yodada
Formula:
C₆H₉I₂NO MW: 364.953 g/mol |
Número MDL:
MFCD00084483 Núm. CAS: 25655-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...