Labels for BradyJet J5000 lab printer
Proveedor: Brady
148652 148653 148656 148658 148665 148666 148651 148654 148655 148657 148659 148660 148661 148662 148663 148664 148667 148668
BRDY148652EA 630 EUR
BRDY148652 BRDY148653 BRDY148656 BRDY148658 BRDY148665 BRDY148666 BRDY148651 BRDY148654 BRDY148655 BRDY148657 BRDY148659 BRDY148660 BRDY148661 BRDY148662 BRDY148663 BRDY148664 BRDY148667 BRDY148668
Labels for BradyJet J5000 lab printer
Etiquetas
This indoor marking film is an aqueous, inkjet printable, flexible, matte finish, pressure sensitive adhesive product that is designed to be used with the BradyJet™ J5000 colour label printer.
  • Specially-formulated topcoat enhances colours and label appearance
  • Topcoat protects print from smudging and harsh conditions
Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting.