Avantor® Hichrom HI-WAX, GC Columns
Proveedor: Avantor
HI09-05-005-2EA 435 EUR
HI09-05-005-2 HI09-05-005-2.5 HI09-05-005-5 HI09-10-010-10 HI09-10-010-15 HI09-10-010-5 HI09-10-020-10 HI09-10-020-15 HI09-10-020-5 HI09-10-040-10 HI09-10-040-15 HI09-10-040-5 HI09-15-015-10 HI09-15-015-15 HI09-15-015-20 HI09-15-015-30 HI09-15-015-5 HI09-25-025-15G5 HI09-25-025-30G5 HI09-25-025-60G5 HI09-53-100-30G5 HI09-18-018-10 HI09-18-018-20 HI09-18-018-40 HI09-18-018-50 HI09-18-018-60 HI09-18-020-10 HI09-18-020-20 HI09-18-020-40 HI09-18-020-50 HI09-18-020-60 HI09-18-040-10 HI09-18-040-20 HI09-18-040-40 HI09-18-040-50 HI09-18-040-60 HI09-20-010-10 HI09-20-010-20 HI09-20-010-40 HI09-20-010-50 HI09-20-010-60 HI09-20-020-10 HI09-20-020-20 HI09-20-020-40 HI09-20-020-50 HI09-20-020-60 HI09-20-040-10 HI09-20-040-20 HI09-20-040-40 HI09-20-040-50 HI09-20-040-60 HI09-20-100-10 HI09-20-100-20 HI09-20-100-40 HI09-20-100-50 HI09-20-100-60 HI09-25-010-15 HI09-25-010-30 HI09-25-010-50 HI09-25-010-60 HI09-25-025-10 HI09-25-025-105 HI09-25-025-15 HI09-25-025-30 HI09-25-025-50 HI09-25-025-60 HI09-25-050-10 HI09-25-050-105 HI09-25-050-15 HI09-25-050-30 HI09-25-050-50 HI09-25-050-60 HI09-25-100-10 HI09-25-100-105 HI09-25-100-15 HI09-25-100-30 HI09-25-100-50 HI09-25-100-60 HI09-32-010-10 HI09-32-010-105 HI09-32-010-15 HI09-32-010-30 HI09-32-010-50 HI09-32-010-60 HI09-32-025-10 HI09-32-025-105 HI09-32-025-15 HI09-32-025-30 HI09-32-025-50 HI09-32-025-60 HI09-32-050-10 HI09-32-050-105 HI09-32-050-15 HI09-32-050-30 HI09-32-050-50 HI09-32-050-60 HI09-32-100-10 HI09-32-100-105 HI09-32-100-15 HI09-32-100-30 HI09-32-100-50 HI09-32-100-60 HI09-32-150-10 HI09-32-150-105 HI09-32-150-15 HI09-32-150-30 HI09-32-150-50 HI09-32-150-60 HI09-32-300-10 HI09-32-300-105 HI09-32-300-15 HI09-32-300-30 HI09-32-300-50 HI09-32-300-60 HI09-53-010-10 HI09-53-010-15 HI09-53-010-30 HI09-53-025-15 HI09-53-025-30 HI09-53-025-50 HI09-53-025-60 HI09-53-050-10 HI09-53-050-15 HI09-53-050-30 HI09-53-050-50 HI09-53-050-60 HI09-53-100-10 HI09-53-100-15 HI09-53-100-30 HI09-53-100-50 HI09-53-100-60 HI09-53-100-75 HI09-53-150-10 HI09-53-150-15 HI09-53-150-30 HI09-53-150-50 HI09-53-150-60 HI09-53-150-75 HI09-53-200-10 HI09-53-200-15 HI09-53-200-30 HI09-53-200-50 HI09-53-200-60 HI09-53-200-75 HI09-53-250-10 HI09-53-250-15 HI09-53-250-30 HI09-53-250-50 HI09-53-250-60 HI09-53-250-75 HIGCHI12-25-025-30 HIGCHI12-32-050-30 HIGCHI09-20-020-25 HIGCHI093203025/35 HIGCHI09-32-025-25 HIGCHI09-32-100-25
Avantor® Hichrom HI-WAX, GC Columns
Columnas de cromatografía
Very popular PEG high polarity phase for a wide range of applications, including (for example): essential oils, flavours and fragrances also alkanols.
  • Phase composition: Cross-bonded Polyethylene Glycol (PEG)
  • Long lifetime
Max. temperature limits may change depending on the film thickness.