Traffic jackets, Iona™, S431
Proveedor: Portwest
S431NARXL S431NARM S431NARS S431NARXXL S431NARL S431NARXXXL
PRTWS431NARXLEA 108 EUR
PRTWS431NARXL PRTWS431NARM PRTWS431NARS PRTWS431NARXXL PRTWS431NARL PRTWS431NARXXXL
Traffic jackets, Iona™, S431
Ropa de trabajo
These traffic jacket combining a 300D outer shell for complete waterproof protection and detachable polar fleece for optimum warmth.
  • Taped seams
  • Pack away hood
  • Phone pocket
  • Print access
  • Radio loop
  • Colour: Navy
The following rainwear collections offer an outstanding choice of designs and colours, suitable for many end users. The best quality fabrics and high technology construction techniques are used throughout, guaranteeing the ultimate in all weather protection. Tested to withstand the rigours of everyday wear, each garment has been designed andMore Product Information
Order Now
ESPECIFICACIONES
Modelo S431
Environmentally Preferable
Aplicaciones de vestimenta General Purpose,Industrial
Risk area Rain (waterproof)
Género Unisex
Peso 60 g/m² + 170 g/m² + 190 g/m²