High visibility overalls with bib and braces, unlined, S388
Proveedor: Portwest
S388ORRL S388ORRM S388ORRS S388ORRXL S388ORRXXL S388ORRXXXL
PRTWS388ORRLEA 60.8 EUR
PRTWS388ORRL PRTWS388ORRM PRTWS388ORRS PRTWS388ORRXL PRTWS388ORRXXL PRTWS388ORRXXXL
High visibility overalls with bib and braces, unlined, S388
Ropa de trabajo
This single colour bib and brace has the features and benefits of the tried and tested S488 contrast bib and brace in a choice of two colours.
  • Reflective tape for increased visibility
  • Six pockets for ample storage
  • Phone pocket
  • Back patch pocket
  • Double rule pocket
  • Elasticated back panel
  • Adjustable side opening for added flexibility
  • Waterproof and breathable with taped seams to prevent water penetration
Manufactured using 100% polyester, 300D oxford weave with a PU coating, 190 g.
Certificaciones: RIS-3279-TOM ISSUE 1; EN 343:2019 Class 4:3 X; EN ISOMore Product Information
Order Now
ESPECIFICACIONES
Modelo S388
Environmentally Preferable
Aplicaciones de vestimenta Industrial,Outdoor
Risk area Rain (waterproof)
Género Unisex
Sleeve or leg length Chest long
Tela Polyester/PU
Peso 380 g