Coenzyme B12
Proveedor: Thermo Fisher Scientific
464360010
464362500
ACRO464360010EA
576
EUR
ACRO464360010
ACRO464362500
Coenzyme B12
Coenzyme B12
- Bioactive
Formula:
C₇₂H₁₀₀CoN₁₈O₁₇P Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00135609 Núm. CAS: 13870-90-1 |
Resultados test especificaciones
Appearance | Red to brown Powder |
Infrared spectrum | Conforms |
Water | ≤12.0 % |
Solubility | (20 mg/ml in water) Clear light to dark red solution |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...