PffBT4T-2DT, Organic Photovoltaics (OPV) reagent
Proveedor: Thermo Fisher Scientific
H66014.MD
H66014.ME
H66014.MC
H66014.MDEA
1380
EUR
H66014.MD
H66014.ME
H66014.MC
PffBT4T-2DT, Organic Photovoltaics (OPV) reagent
PffBT4T-2DT
Formula:
(C₆₆H₉₆N₂S₅F₂)n |
Núm. CAS:
1430201-60-7 |
Resultados test especificaciones
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,65 ± 0,02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...