IEICO-4F, Organic Photovoltaics (OPV) reagent
Proveedor: Thermo Fisher Scientific
H66546.MC
H66546.MD
H66546.ME
H66546.MCEA
709
EUR
H66546.MC
H66546.MD
H66546.ME
IEICO-4F, Organic Photovoltaics (OPV) reagent
Formula:
C₁₁₄H₁₁₄F₄N₄O₄S₄ |
Núm. CAS:
2089044-02-8 |
Resultados test especificaciones
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,28 ± 0,02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...