Chlorophyll A, Sigma-Aldrich®
Proveedor: SIGMA ALDRICH MICROSCOPY
C6144-1MG
C6144-5MG
C6144-1MGEA
260
EUR
C6144-1MG
C6144-5MG
Chlorophyll A, Sigma-Aldrich®
Chlorophyll A
Chlorophyll a has been used as a standard for the calibration of fluorometer.
Formula:
C₅₅H₇₂MgN₄O₅ MW: 893.49 g/mol Pto. de fusión: 150…153 °C Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00079050 Núm. CAS: 479-61-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...