Naphthol AS-MX phosphate disodium salt, Sigma-Aldrich®
Proveedor: SIGMA ALDRICH MICROSCOPY
M-Clarity™ quality level = MQ200
N5000-100MG
N5000-1G
N5000-500MG
N5000-100MGEA
51.4
EUR
N5000-100MG
N5000-1G
N5000-500MG
Naphthol AS-MX phosphate disodium salt, Sigma-Aldrich®
Naphthol AS-MX phosphate disodium salt
Naphthol AS-MX phosphate disodium salt,phosphatase substrate
Formula:
C₁₉H₁₆NNa₂O₅P MW: 415.29 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00069549 Núm. CAS: 96189-12-7 |
|
Resultados test especificaciones
Appearance (Colour) | White to Off-White |
Appearance (Form) | Powder |
Solubility (Colour) | Faint Yellow |
Solubility (Turbidity) 100 mg/ml in H2O |
Clear to Slightly Hazy |
Infrared spectrum | Conforms to Structure |
Water (by Karl Fischer) (10 mol/mol) |
≤31 % |
Carbon (anhydrous) | 53.5 - 56.6 % |
Nitrogen (anhydrous) | 3.0 - 3.6 % |
Phosphorus (P) anhydrous | 7.1 - 7.7 % |
Sodium (Na) anhydrous | 10.0 - 12.0 % |
Purity (HPLC) | ≥99 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...