Oil Blue N 96% (dye content), Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY
Atención

Sinónimos: Solvent Blue 14

M-Clarity™ quality level = MQ200

391557-25G 391557-5G
391557-25GEA 313 EUR
391557-25G 391557-5G
Oil Blue N 96% (dye content), Sigma-Aldrich®
Oil Blue N

Oil Blue N, Dye content 96% (Solvent Blue 14).


In animal tissues, fatty substances are stained dark blue by oil blue N when supersaturated solutions in dilute isopropanol are used.

Formula: C₂₄H₃N₂O₂
Pto. de fusión: 112,..114 °C (lit.)
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00001200
Núm. CAS: 2646-15-3
EINECS: 220-161-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=573783cec7b04e5eab7eab33f8542dacdecaec93&vid=573783cec7b04e5eab7eab33f8542dacdecaec93&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance (Colour) Purple to Very Dark Purple and Black
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength, C = 0.01 g/L; Ethanol 641 - 647 nm
Extinction Coefficient ≥13000
Wavelength 593 - 599 nm
Carbon 72.7 - 79.6%
Nitrogen 7.1 - 7.7%
TLC Consistent with Past Lots
Solubility (Turbidity), C = 1% in Acetone Turbid
Solubility (Colour) Dark Blue to Very Dark Blue


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR