4-Phenoxyphthalonitrile ≥98%, Sigma-Aldrich®
Proveedor: SIGMA ALDRICH MICROSCOPY
|
Atención
|
411930-1G
411930-5G
411930-1GEA
127
EUR
411930-1G
411930-5G
4-Phenoxyphthalonitrile ≥98%, Sigma-Aldrich®
4-Phenoxyphthalonitrile
4-Phenoxyphthalonitrile,98%
Formula:
C₁₄H₈N₂O Pto. de fusión: 98,..100 °C (lit.) Temperatura de almacenaje: Room Temperature |
Número MDL:
MFCD00187787 Núm. CAS: 38791-62-7 |
Resultados test especificaciones
Appearance (Colour) | White to Light Green |
Appearance (Form): Powder, Crystals, Crystalline Powder and/or Chunks | Conforms to Requirements |
Infrared spectrum | Conforms to Structure |
Carbon | 73.7 - 78.3% |
Nitrogen | 12.4 - 13.0% |
Purity (GC) | ≥97.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...