Poly(caprolactone), M.W. ~80000
Proveedor: Thermo Fisher Scientific
178301000
178305000
ACRO178301000EA
78.3
EUR
ACRO178301000
ACRO178305000
Poly(caprolactone), M.W. ~80000
Poly(caprolactone)
Formula:
(C₆H₁₀O₂)n Pto. de fusión: 60 °C |
Número MDL:
MFCD00084404 Núm. CAS: 24980-41-4 |
Resultados test especificaciones
Appearance | White to almost white Beads |
Infrared spectrum | Conforms |
Water | ≤0.5 % |
Molecular weight | 80000 to 85000 g/mol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...