2,3,4-Tri-O-acetyl-α-L-arabinopyranosyl bromide ≥98%
Proveedor: Apollo Scientific
BICL4259-1G
BICL4259-2G
BICL4259-5G
APOSBICL4259-1GEA
393
EUR
APOSBICL4259-1G
APOSBICL4259-2G
APOSBICL4259-5G
2,3,4-Tri-O-acetyl-α-L-arabinopyranosyl bromide ≥98%
2,3,4-Tri-O-acetyl-α-L-arabinopyranosyl bromide
Formula:
C₁₁H₁₅BrO₇ MW: 339,137 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00057497 Núm. CAS: 75247-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...