(R)-α-methyl-phenylglycine
Proveedor: Apollo Scientific
Sinónimos:
N,(R)-α-methyl-phenylglycine
BICR128-1G
BICR128-5G
APOSBICR128-1GEA
429
EUR
APOSBICR128-1G
APOSBICR128-5G
(R)-α-methyl-phenylglycine
(R)-α-methyl-phenylglycine
Formula:
C₉H₁₁NO₂ MW: 165,189 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD07371948 Núm. CAS: 29738-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...