Ethyl amino(cyclopropyl)acetate hydrochloride 95%

Proveedor: Apollo Scientific
Atención

OR015017-1G OR015017-250MG
APOSOR015017-1GEA 835 EUR
APOSOR015017-1G APOSOR015017-250MG
Ethyl amino(cyclopropyl)acetate hydrochloride 95%
Ethyl amino(cyclopropyl)acetate hydrochloride
Formula: C₇H₁₃NO₂·ClH
Temperatura de almacenaje: Ambiente
Número MDL: MFCD18483259
Núm. CAS: 1311313-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR